Learn More
Selleck Chemical LLC Methacholine chloride S3684-25mg
SDP
Catalog No. 503108757
Encompass_Preferred
Methacholine (Mecholyl Acetyl- -methylcholine) is a synthetic choline ester that acts as a non-selective muscarinic receptor agonist in the parasympathetic nervous system but has little effects on the nicotinic receptors
Catalog No. 50-310-8757 Supplier Selleck Chemical LLC Supplier No. S368425MG
Methacholine (Mecholyl Acetyl- -methylcholine) is a synthetic choline ester that acts as a non-selective muscarinic receptor agonist in the parasympathetic nervous system but has little effects on the nicotinic receptors