Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacholine Chloride, High Purity, Spectrum™ Chemical
SDP

Catalog No. 18621037 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-037 1 g
18-621-038 25 g
18-621-039 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-621-037 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M15501GM
Add to Cart
Add to Cart

Synonym: Acetyl-Beta-Methylcholine Chloride, Provocholine, Mecholyl Chloride

Methacholine Chloride, High Purity, a synthetic choline ester, is used to diagnose bronchial hyper reactivity. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 62-51-1

Specifications

Packaging Amber Bottle
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.