Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrolein, 90%, stabilized

Catalog No. AC292835000
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292835000 500 mL Glass bottle
AC292830100 10 mL Glass bottle
AC292830500 50 mL Glass bottle
AC292831000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC292835000 Supplier Thermo Scientific Chemicals Supplier No. 292835000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Methacrolein, 90%, C4H6O, CAS Number-78-85-3, 2-methyl-2-propenal, 2-methylpropenal, 2-methylacrolein, 2-methylenepropanal, 2-propenal, 2-methyl, isobutenal, methacrylaldehyde, methacrylic aldehyde, methylacrylaldehyde, methacrolein, 50mL, 88% min. (GC), 01, III, 2981, 69.0 deg.C | CAS: 78-85-3 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-85-3
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00006974
InChI Key STNJBCKSHOAVAJ-UHFFFAOYSA-N
Synonym methacrolein, methacrylaldehyde, 2-propenal, 2-methyl, isobutenal, 2-methylacrolein, 2-methylpropenal, 2-methyl-2-propenal, methylacrylaldehyde, 2-methylenepropanal, methacrylic aldehyde
PubChem CID 6562
IUPAC Name 2-methylprop-2-enal
SMILES CC(=C)C=O

Specifications

Melting Point -81.0°C
Color Colorless to Yellow
Density 0.8470g/mL
Boiling Point 69.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula H2C=C(CH3)CHO
Refractive Index 1.4145
Quantity 500 mL
Beilstein 01, III, 2981
Specific Gravity 0.847
Solubility Information Solubility in water: 6g/100mL (20°C).
Stabilizer 0.1% hydroquinone
Formula Weight 70.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methacrolein, 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Fatal if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-150-1
RTECSNumber OZ2625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.