Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methacrylamide (2-methylacrylamide) | 79-39-0 | MFCD00008018 | 99.9% | 85.10 g/mol | C4H7NO | 100 G
SDP

Catalog No. 5000300090
Encompass_Preferred

Methacrylamide (2-methylacrylamide) is an acrylic amide monomer used as a reagent and building block in polymer synthesis, hydrogel formation, and materials research. Supplied for research use, it is characterized for purity and accompanied by safety and quality documentation.

  • Molecular formula C4H7NO and molecular weight 85.10 g/mol.
  • High purity (99.9%) for reliable experimental results.
  • Commonly used as a monomer in polymer and hydrogel synthesis.
  • Soluble in common organic solvents such as DMSO for easy handling.
  • Safety data sheet (SDS) and certificate of analysis (COA) available for verification.

Catalog No. 50-003-00090 Supplier Medchemexpress LLC Supplier No. HYW020027100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methacrylamide (2-methylacrylamide) is an acrylic amide monomer used as a reagent and building block in polymer synthesis, hydrogel formation, and materials research. Supplied for research use, it is characterized for purity and accompanied by safety and quality documentation.

  • Molecular formula C4H7NO and molecular weight 85.10 g/mol.
  • High purity (99.9%) for reliable experimental results.
  • Commonly used as a monomer in polymer and hydrogel synthesis.
  • Soluble in common organic solvents such as DMSO for easy handling.
  • Safety data sheet (SDS) and certificate of analysis (COA) available for verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.