Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI Methacrylamide, >98.0% Purity, Powder/Crystal, 25 g

Catalog No. 502781310
Encompass_Preferred

Methacrylamide is a non-heterocyclic amide building block used in chemical synthesis. This compound appears as a white to almost white powder or crystal.

  • Purity: ≥98.0% (GC)
  • Melting point: 109°C to 113°C
  • Molecular formula: C4H7NO
  • Molecular weight: 85.11
  • Physical state: Solid
  • Storage: Room temperature (recommended in a cool and dark place, <15°C)

Catalog No. 50-278-1310 Supplier Grainger Supplier No. 19KR90
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methacrylamide is a non-heterocyclic amide building block used in chemical synthesis. This compound appears as a white to almost white powder or crystal.

  • Purity: ≥98.0% (GC)
  • Melting point: 109°C to 113°C
  • Molecular formula: C4H7NO
  • Molecular weight: 85.11
  • Physical state: Solid
  • Storage: Room temperature (recommended in a cool and dark place, <15°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.