Learn More
Medchemexpress LLC Methacrylamide | 79-39-0 | MFCD00008018 | 99.9% | 85.10 g/mol | C4H7NO | 250 G
SDP
Catalog No. 5000302703
Encompass_Preferred
Methacrylamide is a high-purity monomeric amide (C4H7NO, CAS 79-39-0) used as a building block in polymer synthesis and as a pharmaceutical excipient. As a reactive vinyl amide, it is commonly used in copolymer formation, materials research, and formulation studies where controlled polymerization and functionalization are required.
  • High purity suitable for research and synthesis.
  • Reactive vinyl group enables free-radical polymerization.
  • Useful intermediate for functional and copolymer synthesis.
  • Solid form with established storage recommendations for stability.
  • Applicable to material science, polymer chemistry, and formulation development.
Catalog No. 50-003-02703 Supplier Medchemexpress LLC Supplier No. 250GHYW020027
Methacrylamide is a high-purity monomeric amide (C4H7NO, CAS 79-39-0) used as a building block in polymer synthesis and as a pharmaceutical excipient. As a reactive vinyl amide, it is commonly used in copolymer formation, materials research, and formulation studies where controlled polymerization and functionalization are required.
  • High purity suitable for research and synthesis.
  • Reactive vinyl group enables free-radical polymerization.
  • Useful intermediate for functional and copolymer synthesis.
  • Solid form with established storage recommendations for stability.
  • Applicable to material science, polymer chemistry, and formulation development.