Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methacrylamide | 79-39-0 | MFCD00008018 | 99.9% | 85.10 g/mol | C4H7NO | 500 G
SDP

Catalog No. 5000301814
Encompass_Preferred

Methacrylamide is a high-purity amide monomer for chemical synthesis, polymerization, and use as a pharmaceutical excipient. Supplied as a solid with accompanying analytical documentation, it is intended for research and manufacturing workflows that require a stable, well-characterized monomer.

  • High purity: 99.90% (analytical grade)
  • Physical form: white to off-white solid, easy to handle
  • Molecular formula: C4H7NO; molecular weight: 85.10 g/mol
  • Suitable for polymerization and monomer synthesis
  • Documentation: COA and SDS available for quality and safety verification
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years)

Catalog No. 50-003-01814 Supplier Medchemexpress LLC Supplier No. HYW020027500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methacrylamide is a high-purity amide monomer for chemical synthesis, polymerization, and use as a pharmaceutical excipient. Supplied as a solid with accompanying analytical documentation, it is intended for research and manufacturing workflows that require a stable, well-characterized monomer.

  • High purity: 99.90% (analytical grade)
  • Physical form: white to off-white solid, easy to handle
  • Molecular formula: C4H7NO; molecular weight: 85.10 g/mol
  • Suitable for polymerization and monomer synthesis
  • Documentation: COA and SDS available for quality and safety verification
  • Storage: powder stable at -20°C (3 years) or 4°C (2 years)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.