Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylic acid, 99%, stab. with 100-250ppm hydroquinone or 4-methoxyphenol

Catalog No. AAA115430P
Change view
Click to view available options
Quantity:
500 g
2000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA115430P 2000 g
AAA1154336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA115430P Supplier Thermo Scientific Chemicals Supplier No. A115430P
Add to Cart

CAS: 79-41-4 | C4H6O2 | 86.09 g/mol

Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylic acid is used as a monomer to prepare polymers. It acts a precursor for the preparation methyl methacrylate, N-substituted methacryl amides, methacrylamide, N-substituted methacryl amides and acryloyl chloride. It enhances the adhesion of surface coatings and adhesives. It acts as auxiliaries for the textile industry. It is involved in the preparation of poly(methacrylic acid)/montmorillonite nanocomposites, which finds application in the leather tanning process.

Solubility
Miscible with warm water, chloroform, ethanol, ether and organic solvents.

Notes
Incompatible with amines, strong bases, strong acids, oxidizing agents and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-41-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002651
InChI Key CERQOIWHTDAKMF-UHFFFAOYSA-N
Synonym methacrylic acid, 2-methylacrylic acid, 2-propenoic acid, 2-methyl, methylacrylic acid, 2-methylpropenoic acid, 2-methyl-2-propenoic acid, alpha-methacrylic acid, alpha-methylacrylic acid, 2-methylenepropionic acid, acrylic acid, 2-methyl
PubChem CID 4093
ChEBI CHEBI:25219
IUPAC Name 2-methylprop-2-enoic acid

Specifications

Melting Point 16°C
Density 1.015
pH 2.0 to 2.2
Boiling Point 163°C
Flash Point 65°C
Odor Acrid
Refractive Index 1.431
Quantity 2000 g
UN Number UN2531
Beilstein 1719937
Sensitivity Light and moisture sensitive
Merck Index 14,5941
Solubility Information Miscible with warm water,chloroform,ethanol,ether and organic solvents.
SMILES CC(=C)C(O)=O
Formula Weight 86.09
Percent Purity 99%
Chemical Name or Material Methacrylic acid, Stabilized with 100 to 250 ppm hydroquinone or 4-methoxyphenol
Show More Show Less
Hazard Category H227-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: METHACRYLIC ACID, STABILIZED
EINECSNumber 201-204-4
RTECSNumber OZ2975000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.