Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Methacrylic Acid, Glacial 450 ML
SDP

Catalog No. 5027156
Encompass_Preferred

(2-Methylpropenoic Acid)
CAS #79-41-4 CH₂:C(CH₃)COOH F.W. 86.09
Assay: >98%

Stab. with 0.25% Hydroquinone monomethyl ether

Catalog No. 50-271-56 Supplier Electron Microscopy Sciences Supplier No. 18500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(2-Methylpropenoic Acid)
CAS #79-41-4 CH₂:C(CH₃)COOH F.W. 86.09
Assay: >98%

Stab. with 0.25% Hydroquinone monomethyl ether

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.