Learn More
Medchemexpress LLC Methacrylic acid N-h 1g
SDP
Catalog No. 5000206461
Encompass_Preferred
Methacrylic acid N-hydroxysuccinimide ester (NHS-MA) ios used as a monomer to prepare degradable amphiphilic block copolymer microparticles via RAFT polymerization for low pH-triggered drug delivery[1]
Catalog No. 50-002-06461 Supplier Medchemexpress LLC Supplier No. HYW1054231G
Add to Cart
Add to Cart
Methacrylic acid N-hydroxysuccinimide ester (NHS-MA) ios used as a monomer to prepare degradable amphiphilic block copolymer microparticles via RAFT polymerization for low pH-triggered drug delivery[1]