Learn More
Medchemexpress LLC Methacrylic acid N-h 250mg
SDP
Catalog No. 5000205846
Encompass_Preferred
Methacrylic acid N-hydroxysuccinimide ester (NHS-MA) ios used as a monomer to prepare degradable amphiphilic block copolymer microparticles via RAFT polymerization for low pH-triggered drug delivery[1]
Catalog No. 50-002-05846 Supplier Medchemexpress LLC Supplier No. 250MGHYW105423
Methacrylic acid N-hydroxysuccinimide ester (NHS-MA) ios used as a monomer to prepare degradable amphiphilic block copolymer microparticles via RAFT polymerization for low pH-triggered drug delivery[1]