Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylic Acid, Spectrum™ Chemical
SDP

Catalog No. 18621095 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-095 100 mL
18-621-096 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-621-095 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M2131100ML
Add to Cart
Add to Cart

Synonym: 2-Methylacrylic Acid, Polymethacrylic Acid, 2-Methylpropenoic Acid, Alpha-Methylacrylic Acid

Methacrylic Acid is used as a building block to make a wide range of polymer-based products like binders in paints and coatings, water-soluble polymers and interlevant in glass. Ungraded products supplied by Spectrum™ are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 79-41-4

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.