Learn More
Medchemexpress LLC Methacrylic anhydride | 760-93-0 | MFCD00008586 | 98.8% | 154.17 g/mol | C8H10O3 | 1 G

Supplier: Medchemexpress LLC HYW0173301G
Methacrylic anhydride is a reactive anhydride used to introduce methacryloyl groups into molecules and polymers for subsequent free-radical polymerization and crosslinking. It is intended for laboratory research use, supplied in small pack sizes, and supported by quality and safety documentation for handling and storage.
- Reactive reagent for methacrylation of proteins, polymers, and small molecules.
- Suitable for free-radical polymerization and photo-crosslinking applications.
- High purity (98.8%) for reliable reaction performance.
- Available in small research pack sizes, including 1 g.
- Certificate of Analysis and Safety Data Sheet available for quality and safety reference.
- Storage guidance: pure form at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.