Learn More
Medchemexpress LLC Methacrylic anhydride | 760-93-0 | MFCD00008586 | 98.8% | 154.17 g·mol⁻¹ | C8H10O3 | 100 MG
SDP
Catalog No. 5000300932
Encompass_Preferred
Methacrylic anhydride is a reactive methacrylate derivative used to introduce methacrylate functionality onto polymers, biomolecules, and surfaces. It functions as an acylating reagent in polymer synthesis and surface modification, and is commonly used in free-radical polymerization and biochemical reagent preparation. Handle and store according to safety data recommendations.
  • Reactive methacrylate monomer for acylation and methacrylation of polymers and biomolecules.
  • Enables free-radical polymerization to introduce polymerizable groups.
  • High purity suitable for research and synthesis applications.
  • Available as neat material and as solution formats for ease of use.
  • Stable under recommended storage conditions; follow SDS for handling.
Catalog No. 50-003-00932 Supplier Medchemexpress LLC Supplier No. HYW017330100MG
Methacrylic anhydride is a reactive methacrylate derivative used to introduce methacrylate functionality onto polymers, biomolecules, and surfaces. It functions as an acylating reagent in polymer synthesis and surface modification, and is commonly used in free-radical polymerization and biochemical reagent preparation. Handle and store according to safety data recommendations.
  • Reactive methacrylate monomer for acylation and methacrylation of polymers and biomolecules.
  • Enables free-radical polymerization to introduce polymerizable groups.
  • High purity suitable for research and synthesis applications.
  • Available as neat material and as solution formats for ease of use.
  • Stable under recommended storage conditions; follow SDS for handling.