Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol

Catalog No. AAL1435718
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1435718 50 g
AAL1435730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1435718 Supplier Thermo Scientific Chemicals Supplier No. L1435718
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 760-93-0 | C8H10O3 | 154.17 g/mol

Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.

Solubility
Miscible with ethanol and ether.

Notes
Moisture and light sensitive. Store in a cool place. Incompatible with acids, strong oxidizing agents, alcohols, amines and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 760-93-0
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008586
InChI Key DCUFMVPCXCSVNP-UHFFFAOYSA-N
Synonym methacrylic anhydride, methacrylic acid anhydride, methacryloyl anhydride, 2-propenoic acid, 2-methyl-, anhydride, 2-methyl-2-propenoic acid anhydride, unii-n2rjr03340, 2-methylacrylic anhydride, 2-propenoic acid, 2-methyl-, 1,1'-anhydride, dimethacrylic anhydride, bismethacrylic anhydride
PubChem CID 12974
IUPAC Name 2-methylprop-2-enoyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC(=O)C(C)=C

Specifications

Melting Point -109°C
Density 1.033
Boiling Point 197°C
Flash Point 84°C (183°F)
Refractive Index 1.454
Quantity 50 g
UN Number UN3265
Beilstein 1761982
Sensitivity Light and moisture sensitive
Solubility Information Miscible with ethanol and ether.
Formula Weight 154.17
Percent Purity 94%
Chemical Name or Material Methacrylic anhydride, stabilized with ≈0.2% 2,4-dimethyl-6-tert-butylphenol
Show More Show Less
Hazard Category H227-H302+H332-H315-H317-H318-H335
Hazard Statement GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-084-8
RTECSNumber OZ5700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.