Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methacryloyl chloride

Catalog No. 5232161L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-321-61L 1 L
52-321-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-321-61L Supplier Sigma Aldrich Supplier No. 5232161L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methacryloyl chloride is the clear to slightly colored acid chloride of methacrylic acid. Monomethyl ether hydroquinone is added as a stabilizer to prevent auto polymerization. Reaction with diisopropyl ether in presence of metal salts is rigorous.Methacryloyl chloride is used in the manufacture of polymers:Monomer 2-methacrylamido-caprolactam was prepared by reacting methacryloyl chloride with racemic a-amino-e-caprolactam.Functional monomer was prepared by reacting methacryloyl chloride and L-histidine.A series of amide monomers were synthesized by amidation of methacryloyl chloride with amines and grafted onto commercial poly(ether sulfone) (PES) membranes using irradiation from atmospheric pressure plasma (APP).Reaction of methacryloyl chloride with the hydroxyl groups on the surfaces of HEMA/NVP microspheres was performed, leading to the introduction of polymerisable double bonds onto the surfaces of microspheres.Star-shaped poly(d,l-lactide) oligomers with 2, 3 and 6 arms were synthesised, end-functionalised with methacryloyl chloride and photo-crosslinked in the presence of ethyl lactate as a non-reactive diluent.

Specifications

CAS 920-46-7
Density 1.07 g/mL (at 25°C)
Boiling Point 95°C to 96°C
Molecular Formula C4H5ClO
Refractive Index n20/D 1.442 (literature)
Linear Formula CH2=C(CH3)COCl
Quantity 1 L
MDL Number MFCD00000716
Synonym alpha-Methylacryloyl chloride; 2-Methyl-2-propenoyl chloride; 2-Methylacryloyl chloride; 2-Methylpropenoic acid chloride; Methacrylic acid chloride; Methacrylic chloride
Molecular Weight (g/mol) 104.53
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.