Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methacryloyl chloride, 95%, contains 200 ppm MEHQ as stabilizer, AcroSeal™, Thermo Scientific™

Catalog No. AC168660250
Change view
Click to view available options
Quantity:
25 g
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168660250 25 g AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC168660250 Supplier Thermo Scientific Chemicals Supplier No. 168660250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-46-7
InChI Key VHRYZQNGTZXDNX-UHFFFAOYSA-N
Synonym methacryloyl chloride, methacryloylchloride, methacrylyl chloride, 2-methyl-2-propenoyl chloride, methacryl chloride, methacrylic chloride, methacrylic acid chloride, 2-propenoyl chloride, 2-methyl, 2-methylpropenoyl chloride, methylacryloyl chloride
PubChem CID 13528
IUPAC Name 2-methylprop-2-enoyl chloride
SMILES CC(=C)C(=O)Cl

Specifications

Density 1.0760g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging AcroSeal™ Glass Bottle
Molecular Formula C4H5ClO
Linear Formula H2C=C(CH3)COCl
Quantity 25 g
Stabilizer 200ppm MEHQ max.
Molecular Weight (g/mol) 104.533
Formula Weight 104.54
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Methacryloyl chloride, 97%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
Wear respiratory protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-058-9
RTECSNumber OZ5791000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.