Learn More
Medchemexpress LLC Methacycline hydrochloride | 3963-95-9 | 99.3% | 200 MG
SDP
Catalog No. 5000407745
Encompass_Preferred
Methacycline hydrochloride is a tetracycline antibiotic that inhibits bacterial protein synthesis. It is also a potent epithelial-mesenchymal transition (EMT) inhibitor, blocking EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. This compound is an antimicrobial and has potential for pulmonary fibrosis.
  • Inhibits bacterial protein synthesis
  • Acts as a potent epithelial-mesenchymal transition (EMT) inhibitor
  • Blocks EMT in vitro and fibrogenesis in vivo
  • Demonstrates antimicrobial properties
  • Shows potential for pulmonary fibrosis treatment
  • Inhibits A549 EMT with an IC50 of approximately 5 μM
  • Attenuates Bleomycin-induced canonical EMT markers
  • Improves survival at Day 17 in vivo studies
Catalog No. 50-004-07745 Supplier Medchemexpress LLC Supplier No. HYB0449200MG
Methacycline hydrochloride is a tetracycline antibiotic that inhibits bacterial protein synthesis. It is also a potent epithelial-mesenchymal transition (EMT) inhibitor, blocking EMT in vitro and fibrogenesis in vivo without directly affecting TGF-β1 Smad signaling. This compound is an antimicrobial and has potential for pulmonary fibrosis.
  • Inhibits bacterial protein synthesis
  • Acts as a potent epithelial-mesenchymal transition (EMT) inhibitor
  • Blocks EMT in vitro and fibrogenesis in vivo
  • Demonstrates antimicrobial properties
  • Shows potential for pulmonary fibrosis treatment
  • Inhibits A549 EMT with an IC50 of approximately 5 μM
  • Attenuates Bleomycin-induced canonical EMT markers
  • Improves survival at Day 17 in vivo studies