Learn More
Cliawaived Inc Single Dip Stick Methadone (MTD) Tests
SDP
Catalog No. NC1859225
Encompass_Preferred
(50 Tests)CLIA waived single urine dip stick test for Methadone. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.
Catalog No. NC1859225 Supplier Cliawaived Inc Supplier No. CLIAIDTCII20
(50 Tests)CLIA waived single urine dip stick test for Methadone. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.