Learn More
Medchemexpress LLC Methadp triammonium (adenosine 5'-(α,β-methylene)diphosphate triammonium) | 99.9% | 476.32 | 100 MG
SDP
Catalog No. 5000436333
Encompass_Preferred
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor. It can be used for the research of ATP-adenosine pathway.
  • Acts as a CD73 inhibitor
  • Used for research of the ATP-adenosine pathway
  • Inhibits 5'-AMP induced adenosine formation in A498 cells
Catalog No. 50-004-36333 Supplier Medchemexpress LLC Supplier No. HY112502C100MG
Add to Cart
Add to Cart
MethADP (Adenosine 5'-(α,β-methylene)diphosphate) triammonium is a CD73 inhibitor. It can be used for the research of ATP-adenosine pathway.
  • Acts as a CD73 inhibitor
  • Used for research of the ATP-adenosine pathway
  • Inhibits 5'-AMP induced adenosine formation in A498 cells