Learn More
Strem, An Ascensus Company CAS# 12145-60-7. 1g. Methallylnickel chloride dimer. MFCD04038737
SDP
Supplier:  Strem, An Ascensus Company 2809001G
Encompass
CAS# 12145-60-7. 1g. Methallylnickel chloride dimer. MFCD04038737. Molecular Weight: 298.49. Molecular Formula: [C4H7NiCl]2. Color/form: orange-brown xtl. Strem# 28-0900. http://www.strem.com/catalog/v/28-0900/
Catalog No. 50-901-11242