Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methallyltri-n-butyltin, 98%

Catalog No. AAL2018603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2018603 1 g
AAL2018606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2018603 Supplier Thermo Scientific Chemicals Supplier No. L2018603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 67883-62-9 | C16H34Sn | 345.158 g/mol

Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a methallylating reagent. Also used as a sole curative. It is used as a primary and secondary intermediates in pharmaceuticals.

Solubility
Fully miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67883-62-9
Molecular Formula C16H34Sn
Molecular Weight (g/mol) 345.158
MDL Number MFCD03425867
InChI Key CYIUFVYZGOCGEV-UHFFFAOYSA-N
Synonym methallyltri-n-butyltin, tributyl 2-methylallyl stannane, stannane, tributyl 2-methyl-2-propenyl, tributyl 2-methylprop-2-en-1-yl stannane, methallyltributyltin, 2-methylallyltributyltin, 2-methylallyl tributyltin, methallyltri-n-butylstannane, methallyltri-n-butylin, 2-methylallyl-tributylstannane
PubChem CID 11013379
IUPAC Name tributyl(2-methylprop-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)CC(=C)C

Specifications

Refractive Index 1.4875
Quantity 1 g
UN Number UN2788
Solubility Information Fully miscible with water.
Formula Weight 345.16
Percent Purity 98%
Chemical Name or Material Methallyltri-n-butyltin
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.