Learn More
Cliawaived Inc Single Dip Stick Methamphetamine (mAMP 500) Tests
SDP
Catalog No. NC1859183
Encompass_Preferred
(50 Tests)CLIA waived single urine dip stick test for Methamphetamine. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.
This item is not returnable. View return policy
Catalog No. NC1859183 Supplier Cliawaived Inc Supplier No. CLIAIDTCII16
(50 Tests)CLIA waived single urine dip stick test for Methamphetamine. Read results from 5-60 minutes. New hermetically sealed plastic housing prevent dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.