Learn More
Medchemexpress LLC Methanaminium, 1-carboxy-N,N,N-trimethyl-, chloride (1:1) | 590-46-5 | 153.61 | 10 G
SDP
Catalog No. 5000415363
Encompass_Preferred
Betaine chloride is a natural compound found in many foods and is an active methyl-donor that can maintain normal DNA methylation patterns. It is supplied as a white to off-white solid.
  • Can significantly inhibit HeLa cell proliferation at increasing levels.
  • Prevents hepatic triglyceride accumulation and liver injury when supplemented with alcohol.
  • Improves methylation state of adipose tissue.
  • White to off-white solid appearance.
  • Soluble in water and DMSO.
Catalog No. 50-004-15363 Supplier Medchemexpress LLC Supplier No. HYN073910G
Betaine chloride is a natural compound found in many foods and is an active methyl-donor that can maintain normal DNA methylation patterns. It is supplied as a white to off-white solid.
  • Can significantly inhibit HeLa cell proliferation at increasing levels.
  • Prevents hepatic triglyceride accumulation and liver injury when supplemented with alcohol.
  • Improves methylation state of adipose tissue.
  • White to off-white solid appearance.
  • Soluble in water and DMSO.