Learn More
Medchemexpress LLC Methanaminium, N,N,N-trimethyl-, iodide | 75-58-1 | MFCD00011629 | 98.0% | 201.05 g/mol | C4H12IN | 25 G
SDP
Catalog No. 5000303424
Encompass_Preferred
Tetramethylammonium iodide is a quaternary ammonium salt (CAS 75-58-1) used as a reagent in chemical synthesis and biochemical assays. It is described as a weak surfactant that reduces the surface tension of water and is supplied as a white to off-white powder. Typical analytical values include molecular formula C4H12IN, molecular weight 201.05 g/mol, and typical purity 98.0%.
  • Quaternary ammonium salt for use in chemical synthesis and biochemical assays.
  • Acts as a mild surfactant to reduce surface tension in aqueous solutions.
  • Supplied as a white to off-white powder suitable for laboratory handling.
  • Available in small-scale laboratory pack sizes for research applications.
  • Characterized by molecular formula C4H12IN and molecular weight 201.05 g/mol.
  • Typical purity is 98.0% as shown on the certificate of analysis.
Catalog No. 50-003-03424 Supplier Medchemexpress LLC Supplier No. 25GHYW014861
Tetramethylammonium iodide is a quaternary ammonium salt (CAS 75-58-1) used as a reagent in chemical synthesis and biochemical assays. It is described as a weak surfactant that reduces the surface tension of water and is supplied as a white to off-white powder. Typical analytical values include molecular formula C4H12IN, molecular weight 201.05 g/mol, and typical purity 98.0%.
  • Quaternary ammonium salt for use in chemical synthesis and biochemical assays.
  • Acts as a mild surfactant to reduce surface tension in aqueous solutions.
  • Supplied as a white to off-white powder suitable for laboratory handling.
  • Available in small-scale laboratory pack sizes for research applications.
  • Characterized by molecular formula C4H12IN and molecular weight 201.05 g/mol.
  • Typical purity is 98.0% as shown on the certificate of analysis.