Learn More
Enzo Life Sciences Methanandamide (5mg). CAS: 157182-49-5
SDP
Supplier:  Enzo Life Sciences BMLFA0210005
Encompass_Preferred
Amidase resistant cannabinoid receptor (CB) agonist (CB1: Ki=20nM; CB2: Ki=815nM). The most potent of the series of methyl-anandamides. About 4-fold higher binding affinity for cannabinoid receptor CB1 than anandamide (BML-FA017) in the presence of PMSF. Does also bind to TRPV1 (Ki=4.67µM). Inhibits motor behavior resembling the effectts of THC rather than anadamide.  Alternative name: (R)-(+)-Arachidonyl-1'-hydroxy-2'-propylamide, AM 356, N-(2-Hydroxy-1R-methylethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide. Purity: ≥98% (TLC). Solubility: Soluble in ethanol or DMSO (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0806