Learn More
Medchemexpress LLC Methanesulfonamide, N-[2-chloro-5-[4-[[(1R)-1-phenylethyl]amino]-6-quinazolinyl]-3-pyridinyl]- | 2791417-66-6 | 99.9% | 453.94 | 25 MG
SDP
Catalog No. 5000395426
Encompass_Preferred
MTX-531 is an oral drug that inhibits EGFR and PI3K, demonstrating anti-tumor effects. It also acts as a weak agonist of PPARγ, helping to alleviate hyperglycemia induced by PI3K inhibitors. This compound shows good tolerability in in vivo studies, without causing hyperglycemia, a common side effect of PI3K inhibitors.
  • Inhibits EGFR with an IC50 of 14.7 nM, and PI3K with IC50 values ranging from 1.1 nM to 233 nM.
  • Demonstrates anti-tumor effects, suppressing tumor proliferation and growth.
  • Acts as a weak agonist of PPARγ with an IC50 of 2.5 μM, which helps to alleviate hyperglycemia.
  • Indicates excellent tolerance in mice and the absence of hyperglycemia in vivo.
Catalog No. 50-003-95426 Supplier Medchemexpress LLC Supplier No. 25MGHY162713
MTX-531 is an oral drug that inhibits EGFR and PI3K, demonstrating anti-tumor effects. It also acts as a weak agonist of PPARγ, helping to alleviate hyperglycemia induced by PI3K inhibitors. This compound shows good tolerability in in vivo studies, without causing hyperglycemia, a common side effect of PI3K inhibitors.
  • Inhibits EGFR with an IC50 of 14.7 nM, and PI3K with IC50 values ranging from 1.1 nM to 233 nM.
  • Demonstrates anti-tumor effects, suppressing tumor proliferation and growth.
  • Acts as a weak agonist of PPARγ with an IC50 of 2.5 μM, which helps to alleviate hyperglycemia.
  • Indicates excellent tolerance in mice and the absence of hyperglycemia in vivo.