Learn More
Medchemexpress LLC Methanesulfonamide, N-[3-(1H-indol-4-yl)-5-[(2-methoxy-4-pyridinyl) carbonyl] phenyl]- | 915412-67-8 | >99.94% | C22H19N3O4S | 25 MG
SDP
Catalog No. 5000356524
Encompass_Preferred
LP-261 is a potent, orally active anti-mitotic agent. It demonstrates inhibition of in vitro tubulin polymerization with an EC50 of 3.2 μM. This compound has shown efficacy in inhibiting the growth of human non-small-cell lung tumors (NCI-H522) in vivo, making it a valuable tool for cancer research.
  • Potent anti-mitotic agent.
  • Orally active.
  • Inhibits in vitro tubulin polymerization.
  • Effective against human non-small-cell lung tumors (NCI-H522) in vivo.
  • Useful for cancer research applications.
Catalog No. 50-003-56524 Supplier Medchemexpress LLC Supplier No. 25MGHY14389
LP-261 is a potent, orally active anti-mitotic agent. It demonstrates inhibition of in vitro tubulin polymerization with an EC50 of 3.2 μM. This compound has shown efficacy in inhibiting the growth of human non-small-cell lung tumors (NCI-H522) in vivo, making it a valuable tool for cancer research.
  • Potent anti-mitotic agent.
  • Orally active.
  • Inhibits in vitro tubulin polymerization.
  • Effective against human non-small-cell lung tumors (NCI-H522) in vivo.
  • Useful for cancer research applications.