Learn More
Strem, An Ascensus Company 1g. Methanesulfonato[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [BINAP Palladacycle Gen. 3].
SDP
Supplier:  Strem, An Ascensus Company 4621531G
Encompass
1g. Methanesulfonato[2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [BINAP Palladacycle Gen. 3]. . Molecular Weight: 992.41. Molecular Formula: C57H45NO3P2PdS. Color/form: off-white to light pink pwdr. Strem# 46-2153. http://www.strem.com/catalog/v/46-2153/
Catalog No. NC1111119