Learn More
Strem, An Ascensus Company CAS# 2102544-35-2. 100mg. Methanesulfonato[2-bis(3,5-di(trifluoromethyl)phenylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [JackiePhos Palladacycle Gen. 3]. MFCD22572672
SDP
Supplier:  Strem, An Ascensus Company 460340100MG
Encompass
CAS# 2102544-35-2. 100mg. Methanesulfonato[2-bis(3,5-di(trifluoromethyl)phenylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [JackiePhos Palladacycle Gen. 3]. MFCD22572672. Molecular Weight: 1166.39. Molecular Formula: C52H50F12NO5PPdS. Color/form: white pwdr. Strem# 46-0340. http://www.strem.com/catalog/v/46-0340/
Catalog No. 50-138-5525