Learn More
Strem, An Ascensus Company BrettPhosPalladacycle98% 100mg
SDP
Supplier:  Strem, An Ascensus Company 460322100MG
Encompass
CAS# 1470372-59-8. 100mg. Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-amino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [BrettPhos Palladacycle Gen. 3]. MFCD22572666. Molecular Weight: 906.50. Molecular Formula: C48H66NO5PPdS. Color/form: yellow to beige pwdr. Strem# 46-0322. http://www.strem.com/catalog/v/46-0322/
Catalog No. 50-138-5522