Learn More
Strem, An Ascensus Company CAS# 1651823-59-4. 1g. Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 95% [cataCXium A Palladacycle Gen. 3]. MFCD28144563
SDP
Supplier:  Strem, An Ascensus Company 4602781G
Encompass
CAS# 1651823-59-4. 1g. Methanesulfonato(diadamantyl-n-butylphosphino)-2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 95% [cataCXium A Palladacycle Gen. 3]. MFCD28144563. Molecular Weight: 728.27. Molecular Formula: C37H52NO3PPdS. Color/form: off-white pwdr. Strem# 46-0278. http://www.strem.com/catalog/v/46-0278/
Catalog No. 50-138-5517