Learn More
Strem, An Ascensus Company CAS# 1445085-87-9. 1g. Methanesulfonato[2-(dicyclohexylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) CH2Cl2 adduct, min. 98% [DavePhos Palladacycle Gen. 3]. MFCD28411649
SDP
Supplier:  Strem, An Ascensus Company 4602371G
Encompass
CAS# 1445085-87-9. 1g. Methanesulfonato[2-(dicyclohexylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) CH2Cl2 adduct, min. 98% [DavePhos Palladacycle Gen. 3]. MFCD28411649. Molecular Weight: 763.28. Molecular Formula: C39H49N2O3PPdS. Color/form: white pwdr. Strem# 46-0237. http://www.strem.com/catalog/v/46-0237/
Catalog No. 50-138-5509