Learn More
Strem, An Ascensus Company CAS# 1445086-12-3. 1g. Methanesulfonato(tricyclohexylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 98% [PCy3 Palladacycle Gen. 3]. MFCD28144562
SDP
Supplier:  Strem, An Ascensus Company 4602391G
Encompass
CAS# 1445086-12-3. 1g. Methanesulfonato(tricyclohexylphosphine)(2'-amino-1,1'-biphenyl-2-yl)palladium(II) dichloromethane adduct, min. 98% [PCy3 Palladacycle Gen. 3]. MFCD28144562. Molecular Weight: 650.16. Molecular Formula: C31H46NO3PPdS. Color/form: off-white to beige pwdr. Strem# 46-0239. http://www.strem.com/catalog/v/46-0239/
Catalog No. NC1457152