Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methanesulfonic Acid

Catalog No. 4713565L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-135-65L 5 L
47-135-625L 2.5 L
47-135-625ML 25 mL
47-135-6100ML 100 mL
47-135-6500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 47-135-65L Supplier Sigma Aldrich Supplier No. 4713565L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methanesulfonic Acid

Specifications

CAS 75-75-2
Melting Point 17°C to 19°C
Density 1.481 g/mL at 25°C
Boiling Point 167°C (10 mmHg, lit.)
Molecular Formula CH4O3S
Refractive Index n20/D 1.429
Linear Formula CH3SO3H
MDL Number MFCD00007518
Quantity 5 L
Molecular Weight (g/mol) 96.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.