Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanesulfonic acid, sodium salt | 2386-57-4 | MFCD00064389 | 98.0% | 118.09 g/mol | CH3NaO3S | 1 KG
SDP

Catalog No. 5000300242
Encompass_Preferred

Sodium methanesulfonate is the sodium salt of methanesulfonic acid, supplied as a high-purity, water-soluble white crystalline reagent for laboratory and research use. It is commonly used as a buffer component and additive in biochemical assays and comes with documentation for quality control and safe handling.

  • 98.0% purity
  • Molecular weight 118.09 g/mol
  • Molecular formula CH3NaO3S
  • Water-soluble white crystalline solid
  • Suitable for buffer preparation and biochemical assays
  • Available in 100 g, 500 g, and 1 kg sizes
  • Datasheet, COA, and SDS available for verification

Catalog No. 50-003-00242 Supplier Medchemexpress LLC Supplier No. HYW0343441KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium methanesulfonate is the sodium salt of methanesulfonic acid, supplied as a high-purity, water-soluble white crystalline reagent for laboratory and research use. It is commonly used as a buffer component and additive in biochemical assays and comes with documentation for quality control and safe handling.

  • 98.0% purity
  • Molecular weight 118.09 g/mol
  • Molecular formula CH3NaO3S
  • Water-soluble white crystalline solid
  • Suitable for buffer preparation and biochemical assays
  • Available in 100 g, 500 g, and 1 kg sizes
  • Datasheet, COA, and SDS available for verification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.