Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methanesulfonothioic acid, S-methyl ester | 2949-92-0 | 126.20 | 25 G
SDP

Catalog No. 5000448008
Encompass_Preferred

S-Methyl thiomethanesulfonate (MMTS) is a sulfur-containing volatile organic compound produced by plants and bacteria and is an effective anti-oomycete agent.

  • Effective anti-oomycete agent.
  • Sulfur-containing volatile organic compound.
  • Produced by plants and bacteria.
  • Molecular weight: 126.20.
  • Purity: 99.9%.
  • Appearance: Colorless to light yellow liquid.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-48008 Supplier Medchemexpress LLC Supplier No. 25GHYW004461
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-Methyl thiomethanesulfonate (MMTS) is a sulfur-containing volatile organic compound produced by plants and bacteria and is an effective anti-oomycete agent.

  • Effective anti-oomycete agent.
  • Sulfur-containing volatile organic compound.
  • Produced by plants and bacteria.
  • Molecular weight: 126.20.
  • Purity: 99.9%.
  • Appearance: Colorless to light yellow liquid.
  • Pure form storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.