Learn More
eMolecules​ 124-63-0 | Methanesulfonyl chloride | Oakwood Chemicals | MFCD00007454 | 114.540 | CH3ClO2S | 99.000 | CS(Cl)(=O)=O | 100g | 480109103
Catalog No. NC3918625 Shop All eMolecules Inc. Products
Encompass_Preferred
Methanesulfonyl chloride | Oakwood Chemicals | 124-63-0 | MFCD00007454 | 114.540 | CH3ClO2S | 99.000 | CS(Cl)(=O)=O | 100g | 480109103
Catalog No. NC3918625 Supplier eMolecules​ Supplier No. 480109103
Methanesulfonyl chloride | Oakwood Chemicals | 124-63-0 | MFCD00007454 | 114.540 | CH3ClO2S | 99.000 | CS(Cl)(=O)=O | 100g | 480109103