Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methanesulfonyl Chloride

Catalog No. 642701L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-270-1L 1 L
64-270-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 64-270-1L Supplier Sigma Aldrich Supplier No. 642701L
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methanesulfonyl chloride can be used for the mesylation of primary alcohols to synthesize the corresponding methanesulfonates. It may also be used for the conversion of amines to the corresponding sulfonamides.

Specifications

CAS 124-63-0
Density 1.48 g/mL at 25°C
Boiling Point 60°C (21 mmHg, lit.)
Molecular Formula CH3ClO2S
Linear Formula CH3SO2Cl
Refractive Index n20/D 1.452; n20/D 1.452
MDL Number MFCD00007454
Quantity 1 L
Synonym Methyl sulfonyl chloride; Mesyl chloride
Molecular Weight (g/mol) 114.55 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.