Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich methanesulfonylacetic acid

Catalog No. CDS014245 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014245 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014245 Supplier Sigma Aldrich Supplier No. CDS014245250MG

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-bromo-5-fluorophenol an aryl fluorinated building block.2-bromo-5-fluorophenol may be used for the preparation of reaction intermediates required for the synthesis of polycyclic 5-ht3 and 5-ht4 antagonists
  • It may be used for the the nucleophilic substitution of the 2,2α,4,4α-tetrabromodiphenyl iodonium chloride (iodyl salt)

Specifications

Molecular Formula C3H6O4S
Linear Formula C3H6O4S
Quantity 5 g
MDL Number MFCD00051843
Molecular Weight (g/mol) 138.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.