Learn More
Methanol with 0.1% formic acid, CHROMASOLV™ LC-MS, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Melting Point | -97.68°C |
| CAS | 64-18-6, 67-56-1 |
| Color | Colorless |
| Density | 0.73g/cm3 at 20°C |
| Quantity | 4 x 4 L |
| Explosion Limit | 6% (V) to 36% (V) |
| Flash Point | 11°C (51.8°F) |
| Odor | Odorless |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | P210-P260-P280-P301 + P310 + P330-P308 + P311-P403 + P233 |
| DOTInformation | class 3 - PG 2 - Methanol, solution |
| RTECSNumber | 200-659-6 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.