Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol anhydrous, 67-56-1, MFCD00004595, 1L
Catalog No. 501657108
Encompass
Linear Formula CH3OH, 99.8%, Molecular Weight 32.04, bp 64.7 °C (lit.), mp −98 °C (lit.), Synonym: Methyl alcohol
Catalog No. 50-165-7108 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3224151L
Linear Formula CH3OH, 99.8%, Molecular Weight 32.04, bp 64.7 °C (lit.), mp −98 °C (lit.), Synonym: Methyl alcohol