Learn More
Cell Signaling Technology Methanol, 99.8% Purity, 125 mL
SDP
Catalog No. 502050572
Encompass
This premium methanol is submicron filtered and offers 99.8% purity for use in biological assays. It is commonly utilized as a fixative or permeabilization agent in immunofluorescence and flow cytometry experiments. Please consult the specific antibody datasheet to confirm if methanol fixation or permeabilization is recommended for your application.
  • 99.8% pure and submicron filtered
  • Ideal for use as a fixative or permeabilization agent
  • Stable for 12 months when stored at room temperature
  • Chill at -20°C prior to use
Catalog No. 50-205-0572 Supplier Cell Signaling Technology Supplier No. 13604S
This premium methanol is submicron filtered and offers 99.8% purity for use in biological assays. It is commonly utilized as a fixative or permeabilization agent in immunofluorescence and flow cytometry experiments. Please consult the specific antibody datasheet to confirm if methanol fixation or permeabilization is recommended for your application.
  • 99.8% pure and submicron filtered
  • Ideal for use as a fixative or permeabilization agent
  • Stable for 12 months when stored at room temperature
  • Chill at -20°C prior to use