Learn More
VWR INTERNATIONAL LLC Methanol ≥99.8% for HPLC, 67-56-1, MFCD00004595, 4L
Catalog No. NC1179385
Encompass
Formula: CH₃OH, MW: 32.04 g/mol, Boiling Pt: 64.6 °C (1013 hPa), Melting Pt: –98 °C, Storage Temperature: ambient, Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol.
Catalog No. NC1179385 Supplier VWR INTERNATIONAL LLC Supplier No. 97065064
Formula: CH₃OH, MW: 32.04 g/mol, Boiling Pt: 64.6 °C (1013 hPa), Melting Pt: –98 °C, Storage Temperature: ambient, Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol.