Learn More
Research Products International Corp Methanol, 5 Gal
SDP
Catalog No. 501977473
Encompass_Preferred
Colorless liquid, Flammable, Flash pt. 12°C, b.p. 64-65°C
WARNING! This product can expose you to Methanol, a chemical which is known to the State of California to cause Developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7473 Supplier Research Products International Corp Supplier No. 400032
Colorless liquid, Flammable, Flash pt. 12°C, b.p. 64-65°C
WARNING! This product can expose you to Methanol, a chemical which is known to the State of California to cause Developmental harm. For more information go to www.P65Warnings.ca.gov.