Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Methanol 62% W/W 4l Plastic Bottle

Catalog No. 504400900
Encompass_Preferred

Methanol 62% W/W-Jug 1 Gallon Plastic

Catalog No. 50-440-0900 Supplier LaMarKa Chemical Company Supplier No. 00007564766932436
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methanol 62% W/W-Jug 1 Gallon Plastic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.