Learn More
Sigma Aldrich Fine Chemicals Biosciences Methanol, 67-56-1, MFCD00004595, 1 L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 348601LR
Encompass
Linear Formula: CH3OH, Molecular Weight: 32.04, for HPLC, ≥99.9%, Synonym: Methyl alcohol. Methanol can be used as a mobile phase in reversed-phase liquid chromatography. Methanol (MeOH) has been used in the wholemount immunofluorescence studies of embryonic tissues.
Catalog No. NC0446254