Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Methanol

Catalog No. 01257784 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Methanol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-257-784 Methanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-784 Supplier MilliporeSigma Supplier No. 1047321000
Add to Cart
Add to Cart

Methanol tested for EPA 533 and EPA 537.1 PFAS methods LiChrosolv™.

  • PFAS tested solvents are used for UHPLC-MS and LC-MS instrumental analysis.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methanol
CAS 67-56-1
Quantity 1 L
Packaging Borosilicate Glass Bottle
Boiling Point 64.7°C
Molecular Weight (g/mol) 32.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.