Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019476 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9476 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9476 Supplier MilliporeSigma Supelco Supplier No. 8276210MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methanol is a primary alcohol, which is commercially manufactured from methane. It can also be obtained via pyrolysis of wood. It can find applications in organic synthesis reactions, as a highly polar solvent.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1
Ignition Point 725°F
Vapor Pressure 410 mmHg (50°C)
Vapor Density 1.11 (vs. air)
Density 0.791 g/mL (at 25°C)
Flash Point 9°C
Physical Form Neat
Linear Formula CH3 OH
MDL Number MFCD00004595
Melting Point -98°C
Boiling Point 64.7°C
Synonym Methyl alcohol
Type Analytical Standard
Quantity 10 mL
Formula Weight 32.04
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.