Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Absolute, Vion Biosciences
SCI_ED

Catalog No. S02491 Shop All Aldon Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02491 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.
Catalog No. S02491 Supplier Aldon Corporation Supplier No. VNMM03601L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to Methanol, which is known to the State of California to cause birth defects or other reproductive harm.

This histology grade methanol is an ideal solvent for fixation and dehydration in the histology lab.

  • This histology grade methanol is an ideal solvent for fixation and dehydration in the histology lab.

Chemical Identifiers

Molecular Formula CH3OH
Molecular Weight (g/mol) 32.04 g/mol
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym None.
PubChem CID Data not available.
ChEBI CHEBI:17790
IUPAC Name Methanol
SMILES CO

Specifications

pH Data not available.
Melting Point -98°C
Color Colorless
CAS Max % 100%
Appearance Clear, colorless liquid.
Decomposition Information Data not available.
Density 0.79 g/mL
CAS Min % 100%
Quantity 1 L
CAS 67-56-1
Identification Methanol
Ignition Point 463°C
Odor Pungent odor.
Packaging Natural Plastic Cylinder
Physical Form Data not available.
Product Line Data not available.
Assay Percent Range 100%
Vapor Pressure 96 mm (at 20°C)
Linear Formula CH3OH
UN Number UN1230
Water 0%
Concentration 100%
Solubility Information Complete in water.
Boiling Point 65°C
Percent Purity 100%
Grade Data not available.
Concentration or Composition (by Analyte or Components) 100% Methanol
Chemical Name or Material Methanol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Acute Toxicity, Oral
Hazard Statement Acute Toxicity, Dermal
Precautionary Statement Acute Toxicity, Inhalation
ShelfLife 3 Years
DOTInformation UN1230, Methanol, Domestic 3; International 3; 6.1 (II), Limited Quantity ≥ 1 L
RTECSNumber PC1400000
Recommended Storage Store in a cool, dry, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.